Assay Details
| Gene Symbol: Xpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011728.2 | NP_035858.2 | 4 - 5 | 647 | 85 | |
| GenBank mRNA | AK008962.1 | - | 4 - 5 | 641 | 85 | |
| BC029241.1 | - | 624 | 4489063 | 85 | ||
| BP771655.1 | - | 2 - 3 | 85 | |||
| X74351.1 | - | 4 - 5 | 647 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xpa
Genomic Map