Assay Details
| Gene Symbol: Pigq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011822.4 | NP_035952.2 | 4 - 5 | 1125 | 76 | |
| GenBank mRNA | AB008895.1 | - | 4 - 5 | 1080 | 76 | |
| AK155173.1 | - | 4 - 5 | 1028 | 76 | ||
| AK163680.1 | - | 4 - 5 | 1101 | 76 | ||
| AK169284.1 | - | 4 - 5 | 1108 | 76 | ||
| AK188686.1 | - | 2 - 3 | 184 | 76 | ||
| AK195137.1 | - | 1 - 2 | 54 | 76 | ||
| AK214222.1 | - | 2 - 3 | 76 | |||
| AK218878.1 | - | 3 - 4 | 318 | 76 | ||
| BC003917.1 | - | 2 - 3 | 215 | 76 | ||
| BC014287.1 | - | 4 - 5 | 1098 | 4193343 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigq
Genomic Map