Assay Details
| Gene Symbol: Pigq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291025.1 | NP_001277954.1 | 8 - 9 | 1696 | 62 | |
| NM_011822.4 | NP_035952.2 | 7 - 8 | 1516 | 62 | ||
| GenBank mRNA | AB008895.1 | - | 7 - 8 | 1471 | 62 | |
| AK155173.1 | - | 6 - 7 | 1697 | 62 | ||
| AK163680.1 | - | 7 - 8 | 1492 | 62 | ||
| AK169284.1 | - | 7 - 8 | 1499 | 62 | ||
| AK170431.1 | - | 8 - 9 | 1643 | 62 | ||
| AK213710.1 | - | 1 - 2 | 60 | 62 | ||
| BC003917.1 | - | 5 - 6 | 606 | 62 | ||
| BC014287.1 | - | 7 - 8 | 1489 | 4193343 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigq
Genomic Map