Assay Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163183.1 | NP_001156655.1 | 5 - 6 | 539 | 94 | |
| NM_001163186.1 | NP_001156658.1 | 4 - 5 | 530 | 94 | ||
| NM_013746.3 | NP_038774.1 | 6 - 7 | 686 | 94 | ||
| GenBank mRNA | AF000272.1 | - | 6 - 7 | 657 | 94 | |
| AF100613.1 | - | 4 - 5 | 518 | 94 | ||
| BC024756.1 | - | 6 - 7 | 647 | 5369461 | 94 | |
| CX201870.1 | - | 5 - 6 | 491 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhb1
Genomic Map