Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013786.2 | NP_038814.1 | 3 - 4 | 508 | 73 | |
| GenBank mRNA | AK008212.1 | - | 3 - 4 | 567 | 73 | |
| BC021836.1 | - | 3 - 4 | 508 | 5135142 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b6
Genomic Map