Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013786.2 | NP_038814.1 | 4 - 5 | 759 | 105 | |
| GenBank mRNA | BC021836.1 | - | 4 - 5 | 759 | 5135142 | 105 |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b6
Genomic Map