Assay Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038700.2 | NP_001033789.1 | 2 - 3 | 344 | 75 | |
| NM_001177648.1 | NP_001171119.1 | 2 - 3 | 344 | 75 | ||
| NM_001177649.1 | NP_001171120.1 | 2 - 3 | 344 | 75 | ||
| NM_019406.3 | NP_062279.1 | 2 - 3 | 344 | 75 | ||
| GenBank mRNA | AK122308.1 | - | 2 - 3 | 344 | 75 | |
| AK157135.1 | - | 2 - 3 | 335 | 75 | ||
| AK170497.1 | - | 2 - 3 | 294 | 75 | ||
| AK172100.1 | - | 2 - 3 | 288 | 75 | ||
| BC003867.1 | - | 2 - 3 | 275 | 3490113 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fnbp1
Genomic Map