Assay Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038700.2 | NP_001033789.1 | 4 - 5 | 553 | 72 | |
| NM_001177648.1 | NP_001171119.1 | 4 - 5 | 553 | 72 | ||
| NM_001177649.1 | NP_001171120.1 | 4 - 5 | 553 | 72 | ||
| NM_001177650.1 | NP_001171121.1 | 2 - 3 | 250 | 72 | ||
| NM_019406.3 | NP_062279.1 | 4 - 5 | 553 | 72 | ||
| GenBank mRNA | AK122308.1 | - | 4 - 5 | 553 | 72 | |
| AK143512.1 | - | 2 - 3 | 251 | 72 | ||
| AK157135.1 | - | 4 - 5 | 544 | 72 | ||
| AK170497.1 | - | 4 - 5 | 503 | 72 | ||
| AK172100.1 | - | 4 - 5 | 497 | 72 | ||
| BC003867.1 | - | 4 - 5 | 484 | 3490113 | 72 | |
More Information
Set Membership: |
|

Fnbp1
Genomic Map