Assay Details
| Gene Symbol: Prodh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019546.5 | NP_062419.2 | 2 - 3 | 392 | 65 | |
| GenBank mRNA | AF222851.1 | - | 2 - 3 | 379 | 65 | |
| AK011020.1 | - | 2 - 3 | 383 | 65 | ||
| AK011039.1 | - | 2 - 3 | 392 | 65 | ||
| AK050141.1 | - | 2 - 3 | 393 | 65 | ||
| BC018182.1 | - | 2 - 3 | 378 | 4161894 | 65 | |
| U80019.1 | - | 2 - 3 | 390 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prodh2
Genomic Map