Assay Details
| Gene Symbol: Tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019551.2 | NP_062424.1 | 5 - 6 | 741 | 85 | |
| GenBank mRNA | AJ251328.1 | - | 5 - 6 | 715 | 85 | |
| AK009089.1 | - | 5 - 6 | 715 | 85 | ||
| AK167707.1 | - | 4 - 5 | 537 | 85 | ||
| BC132511.1 | - | 5 - 6 | 704 | 40130788 | 85 | |
| BC132513.1 | - | 5 - 6 | 704 | 40130790 | 85 | |
| BC145334.1 | - | 4 - 5 | 533 | 85 | ||
More Information
Set Membership: |
|

Tdp2
Genomic Map