Assay Details
| Gene Symbol: Ypel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023249.6 | NP_075738.1 | 1 - 2 | 225 | 116 | |
| GenBank mRNA | AF190624.1 | - | 1 - 2 | 141 | 116 | |
| AK006858.1 | - | 1 - 2 | 113 | 116 | ||
| AK076063.1 | - | 1 - 2 | 168 | 116 | ||
| AK133401.1 | - | 1 - 2 | 139 | 116 | ||
| AK139453.1 | - | 1 - 2 | 133 | 116 | ||
| BB649357.1 | - | 1 - 2 | 225 | 116 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF190624.1,AK006858.1,AK076063.1,AK133401.1,AK139453.1,NM_023249.6 transcripts.
Set Membership: |
|

Ypel1
Genomic Map