Assay Details
| Gene Symbol: Serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023475.3 | NP_075964.1 | 4 - 5 | 453 | 80 | |
| GenBank mRNA | AJ245737.1 | - | 456 | 80 | ||
| AJ251200.1 | - | 4 - 5 | 456 | 80 | ||
| AK002313.1 | - | 4 - 5 | 366 | 80 | ||
| AK003827.1 | - | 4 - 5 | 366 | 80 | ||
| AK154770.1 | - | 3 - 4 | 1661 | 80 | ||
| BC024048.1 | - | 4 - 5 | 334 | 80 | ||
| BC046342.1 | - | 4 - 5 | 338 | 6475904 | 80 | |
| BC055431.1 | - | 4 - 5 | 465 | 3673247 | 80 | |
More Information
Set Membership: |
|

Serhl
Genomic Map