Assay Details
| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024242.3 | NP_077204.2 | 10 - 11 | 1467 | 77 | |
| GenBank mRNA | AK017312.1 | - | 10 - 11 | 1162 | 77 | |
| AK152707.1 | - | 10 - 11 | 1216 | 77 | ||
| AK161315.1 | - | 11 - 12 | 1710 | 77 | ||
| BC002158.1 | - | 4 - 5 | 424 | 3485522 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok1
Genomic Map