Assay Details
| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024242.3 | NP_077204.2 | 12 - 13 | 1675 | 62 | |
| GenBank mRNA | AK017312.1 | - | 12 - 13 | 1370 | 62 | |
| AK152707.1 | - | 12 - 13 | 1424 | 62 | ||
| AK161315.1 | - | 13 - 14 | 1918 | 62 | ||
| BC002158.1 | - | 6 - 7 | 632 | 3485522 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok1
Genomic Map