Assay Details
| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002497.1 | - | 1 - 2 | 437 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002497.1 transcripts.
Set Membership: |
|

Wbscr22
Genomic Map