Assay Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025745.3 | NP_080021.3 | 12 - 13 | 1360 | 70 | |
| GenBank mRNA | AK002471.1 | - | 13 - 14 | 1351 | 70 | |
| AK077120.1 | - | 12 - 13 | 1353 | 70 | ||
| AK133874.1 | - | 12 - 13 | 1361 | 70 | ||
| AK162571.1 | - | 2 - 3 | 129 | 70 | ||
| BC018468.1 | - | 12 - 13 | 1370 | 3591901 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erlec1
Genomic Map