Assay Details
| Gene Symbol: Wwp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025830.3 | NP_080106.1 | 19 - 20 | 2176 | 62 | |
| GenBank mRNA | AK004962.2 | - | 19 - 20 | 2192 | 62 | |
| AK088936.1 | - | 19 - 20 | 2155 | 62 | ||
| AK146306.1 | - | 18 - 19 | 2038 | 62 | ||
| AK151548.1 | - | 19 - 20 | 2170 | 62 | ||
| AK159248.1 | - | 10 - 11 | 1220 | 62 | ||
| BC004712.1 | - | 4 - 5 | 496 | 62 | ||
| BC018496.1 | - | 3 - 4 | 1374 | 62 | ||
| BC039921.1 | - | 19 - 20 | 2168 | 4954925 | 62 | |
| BC048184.1 | - | 19 - 20 | 2176 | 6827381 | 62 | |
| HQ896246.1 | - | 10 - 11 | 1273 | 62 | ||
| JN712751.1 | - | 19 - 20 | 2209 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wwp2
Genomic Map