Assay Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025948.2 | NP_080224.1 | 6 - 7 | 837 | 102 | |
| GenBank mRNA | AK003611.1 | - | 4 - 5 | 397 | 102 | |
| AK012276.1 | - | 5 - 6 | 600 | 102 | ||
| BC031521.1 | - | 6 - 7 | 837 | 2655161 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14a
Genomic Map