Assay Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025948.2 | NP_080224.1 | 9 - 10 | 1430 | 79 | |
| GenBank mRNA | AK003611.1 | - | 7 - 8 | 990 | 79 | |
| AK012276.1 | - | 8 - 9 | 1193 | 79 | ||
| BC031521.1 | - | 9 - 10 | 1430 | 2655161 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14a
Genomic Map