Assay Details
| Gene Symbol: Agpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026212.2 | NP_080488.1 | 2 - 3 | 516 | 77 | |
| GenBank mRNA | AK010891.1 | - | 2 - 3 | 325 | 77 | |
| AK090271.1 | - | 2 - 3 | 518 | 77 | ||
| AY072769.1 | - | 2 - 3 | 309 | 77 | ||
| BC125530.1 | - | 2 - 3 | 390 | 40130045 | 77 | |
| BC137840.1 | - | 2 - 3 | 390 | 8860859 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat2
Genomic Map