Assay Details
| Gene Symbol: Agpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026212.2 | NP_080488.1 | 3 - 4 | 706 | 72 | |
| GenBank mRNA | AK010891.1 | - | 3 - 4 | 515 | 72 | |
| AK090271.1 | - | 3 - 4 | 708 | 72 | ||
| AY072769.1 | - | 3 - 4 | 499 | 72 | ||
| BC125530.1 | - | 3 - 4 | 580 | 40130045 | 72 | |
| BC137840.1 | - | 3 - 4 | 580 | 8860859 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat2
Genomic Map