Assay Details
| Gene Symbol: Shisa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026381.5 | NP_080657.1 | 1 - 2 | 147 | 96 | |
| GenBank mRNA | AK018291.1 | - | 1 - 2 | 103 | 96 | |
| AK020877.1 | - | 1 - 2 | 124 | 96 | ||
| AK081060.1 | - | 1 - 2 | 125 | 96 | ||
| BC017600.1 | - | 1 - 2 | 59 | 4487600 | 96 | |
| BC057089.1 | - | 1 - 2 | 99 | 5689110 | 96 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK018291.1,AK020877.1,AK081060.1,BC017600.1,BC057089.1,NM_026381.5 transcripts.
Set Membership: |
|

Shisa5
Genomic Map