Assay Details
| Gene Symbol: Poldip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026389.3 | NP_080665.1 | 5 - 6 | 529 | 106 | |
| GenBank mRNA | AK003702.1 | - | 3 - 4 | 195 | 106 | |
| AK165703.1 | - | 5 - 6 | 561 | 106 | ||
| BC006833.1 | - | 5 - 6 | 529 | 3597851 | 106 | |
| BC012879.1 | - | 5 - 6 | 541 | 4166791 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip2
Genomic Map