Assay Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027570.3 | NP_081846.3 | 8 - 9 | 1098 | 66 | |
| GenBank mRNA | AK014649.1 | - | 8 - 9 | 1104 | 66 | |
| AK029820.1 | - | 1512 | 66 | |||
| AK037996.1 | - | 408 | 66 | |||
| AY092768.1 | - | 8 - 9 | 1103 | 66 | ||
| BC039155.1 | - | 8 - 9 | 1094 | 4242682 | 66 | |
| BC055443.1 | - | 8 - 9 | 1098 | 4235345 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ATP biosynthetic process
oxidation-reduction process
catalytic activity
D-lactate dehydrogenase (cytochrome) activity
protein binding
D-lactate dehydrogenase activity
UDP-N-acetylmuramate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
flavin adenine dinucleotide binding
ATP biosynthetic process
oxidation-reduction process
catalytic activity
D-lactate dehydrogenase (cytochrome) activity
protein binding
D-lactate dehydrogenase activity
UDP-N-acetylmuramate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018697 | BC055443.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018697 | AK029820.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018697 | AK037996.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018697 | NM_027570.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532802 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138781 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018697 | BC055443.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018697 | AK029820.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018697 | AK037996.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018697 | NM_027570.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161454 | BC055443.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161454 | BC039155.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161454 | AK029820.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161454 | AK037996.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P161454 | NM_027570.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300038 | AK014649.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26169352 | - | - |
| Whole Mouse Genome G4122A | A_52_P161454 | BC055443.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P161454 | BC039155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P161454 | AK029820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P161454 | AK037996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P161454 | NM_027570.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300038 | AK014649.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678783 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06678811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26169337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07181143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06807610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06734971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06678785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06678771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06678816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06678783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06825389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06678770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26169384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581869 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428613_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428614_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581865 | - | - |
| Mouse Genome 430 2.0 Array | 1428614_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428614_at | - | - |
| Mouse Genome 430 2.0 Array | 1428613_at | - | - |
| Mouse Genome 430 2.0 Array | 1428613_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52815_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52815_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800022789.mm.1 | NM_027570.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022789.mm.1 | NM_027570.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022789.mm.1 | TC0800002950.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022788.mm.1 | NM_027570.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022788.mm.1 | NM_027570.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022788.mm.1 | TC0800002950.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428613_PM_at | - | - |
| Mouse Expression Set 430 | 1428613_at | - | - |
| Mouse Expression Set 430 | 1428614_PM_at | - | - |
| Mouse Expression Set 430 | 1428614_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428614_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428613_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428613_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428614_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900504 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900504 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665629 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639662 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639662 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665629 | - | - |
| mouse-6 v1.1 (Illumina) | scl0052815.1_84-S | - | - |
| mouse-6 v1.1 (Illumina) | scl052815.2_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120100 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900504 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0052815.1_84-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl052815.2_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120100 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900504 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639662 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2665629 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2825080 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665629 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2825080 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639662 | - | - |

Ldhd
Genomic Map