Assay Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB093224.1 | - | 5 - 6 | 2002 | 81 | |
| AF363690.1 | - | 10 - 11 | 2732 | 81 | ||
| AK004685.1 | - | 1 - 2 | 131 | 81 | ||
| AK147276.1 | - | 10 - 11 | 3365 | 81 | ||
| AK147535.1 | - | 11 - 12 | 5483 | 81 | ||
| BC040463.1 | - | 1 - 2 | 432 | 81 | ||
| BC058665.1 | - | 7 - 8 | 2250 | 81 | ||
More Information
Set Membership: |
|

Cic
Genomic Map