Assay Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB093224.1 | - | 7 - 8 | 2310 | 63 | |
| AF363690.1 | - | 12 - 13 | 3040 | 63 | ||
| AK004685.1 | - | 3 - 4 | 439 | 63 | ||
| AK082152.1 | - | 2 - 3 | 202 | 63 | ||
| AK147276.1 | - | 12 - 13 | 3673 | 63 | ||
| AK147535.1 | - | 13 - 14 | 5791 | 63 | ||
| BC040463.1 | - | 3 - 4 | 740 | 63 | ||
| BC058665.1 | - | 9 - 10 | 2558 | 63 | ||
More Information
Set Membership: |
|

Cic
Genomic Map