| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300015945.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015945.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015945.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015941.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015941.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015941.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015948.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015948.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015948.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015942.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015942.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015942.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015940.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015940.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015940.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015937.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015937.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015937.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015938.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015938.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015938.mm.1 |
TC1300002395.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015939.mm.1 |
NM_028012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015939.mm.1 |
NM_028012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015939.mm.1 |
TC1300002395.mm.1 |
MTA TC |