Assay Details
| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029556.3 | NP_083832.2 | 2 - 3 | 276 | 78 | |
| GenBank mRNA | AF428254.1 | - | 2 - 3 | 249 | 78 | |
| AK002741.1 | - | 2 - 3 | 402 | 78 | ||
| AK009345.1 | - | 2 - 3 | 264 | 78 | ||
| BC023398.1 | - | 2 - 3 | 266 | 5064709 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002741.1 transcripts.
Set Membership: |
|

Clybl
Genomic Map