Assay Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029746.3 | NP_084022.2 | 13 - 14 | 1671 | 91 | |
| GenBank mRNA | AK004441.1 | - | 1 - 2 | 149 | 91 | |
| AK169298.1 | - | 13 - 14 | 1641 | 91 | ||
| AK171726.1 | - | 13 - 14 | 1590 | 91 | ||
| BC011435.1 | - | 13 - 14 | 1611 | 4010102 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cog2
Genomic Map