Assay Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029746.3 | NP_084022.2 | 14 - 15 | 1744 | 83 | |
| GenBank mRNA | AK004441.1 | - | 2 - 3 | 222 | 83 | |
| AK169298.1 | - | 14 - 15 | 1714 | 83 | ||
| AK171726.1 | - | 14 - 15 | 1663 | 83 | ||
| AK178946.1 | - | 2 - 3 | 141 | 83 | ||
| AK213396.1 | - | 1 - 2 | 87 | 83 | ||
| BC011435.1 | - | 14 - 15 | 1684 | 4010102 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cog2
Genomic Map