Assay Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029775.2 | NP_084051.1 | 6 - 7 | 788 | 114 | |
| GenBank mRNA | AK013933.1 | - | 6 - 7 | 806 | 114 | |
| AK014737.1 | - | 6 - 7 | 830 | 114 | ||
| AK134862.1 | - | 835 | 114 | |||
| BC020048.1 | - | 6 - 7 | 824 | 114 | ||
| BC020089.1 | - | 6 - 7 | 788 | 4013755 | 114 | |
| BC049641.1 | - | 977 | 114 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcun1d5
Genomic Map