Assay Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145800.1 | NP_001139272.1 | 6 - 7 | 928 | 92 | |
| NM_033608.3 | NP_291086.2 | 6 - 7 | 994 | 92 | ||
| GenBank mRNA | AF317839.1 | - | 6 - 7 | 994 | 92 | |
| AK078536.1 | - | 6 - 7 | 922 | 92 | ||
| AK145121.1 | - | 6 - 7 | 903 | 92 | ||
| BC034594.1 | - | 6 - 7 | 910 | 92 | ||
| BC040281.1 | - | 6 - 7 | 954 | 5364073 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf9
Genomic Map