Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100013470.mm.1 |
NM_001145800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013470.mm.1 |
NM_001145800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013470.mm.1 |
NM_033608.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013470.mm.1 |
NM_033608.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013470.mm.1 |
TC0100001616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013469.mm.1 |
NM_001145800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013469.mm.1 |
NM_001145800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013469.mm.1 |
NM_033608.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013469.mm.1 |
NM_033608.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013469.mm.1 |
TC0100001616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013468.mm.1 |
NM_001145800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013468.mm.1 |
NM_001145800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013468.mm.1 |
NM_033608.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013468.mm.1 |
NM_033608.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013468.mm.1 |
TC0100001616.mm.1 |
MTA TC |