Assay Details
| Gene Symbol: Eri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285899.1 | NP_001272828.1 | 3 - 4 | 735 | 70 | |
| NM_001285901.1 | NP_001272830.1 | 2 - 3 | 318 | 70 | ||
| NM_080469.4 | NP_536717.2 | 3 - 4 | 721 | 70 | ||
| GenBank mRNA | AK083017.1 | - | 3 - 4 | 592 | 70 | |
| AK144107.1 | - | 3 - 4 | 723 | 70 | ||
| AK169843.1 | - | 1 - 2 | 290 | 70 | ||
| BC021405.1 | - | 1 - 2 | 271 | 70 | ||
| BC096545.1 | - | 3 - 4 | 2868 | 70 | ||
| BC116685.1 | - | 3 - 4 | 568 | 40105869 | 70 | |
| BC117540.1 | - | 3 - 4 | 582 | 40105872 | 70 | |
| BY258104.1 | - | 2 - 3 | 296 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eri3
Genomic Map