Assay Details
| Gene Symbol: Eri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285899.1 | NP_001272828.1 | 6 - 7 | 1006 | 68 | |
| NM_001285901.1 | NP_001272830.1 | 5 - 6 | 589 | 68 | ||
| NM_001285902.1 | NP_001272831.1 | 4 - 5 | 311 | 68 | ||
| NM_080469.4 | NP_536717.2 | 6 - 7 | 992 | 68 | ||
| GenBank mRNA | AK083017.1 | - | 6 - 7 | 863 | 68 | |
| AK144107.1 | - | 6 - 7 | 994 | 68 | ||
| AY029599.1 | - | 3 - 4 | 185 | 68 | ||
| BC021405.1 | - | 4 - 5 | 542 | 68 | ||
| BC079640.1 | - | 2 - 3 | 156 | 68 | ||
| BC096545.1 | - | 6 - 7 | 3139 | 68 | ||
| BC116685.1 | - | 6 - 7 | 839 | 40105869 | 68 | |
| BC117540.1 | - | 6 - 7 | 853 | 40105872 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eri3
Genomic Map