Assay Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242389.1 | NP_001229318.1 | 6 - 7 | 613 | 54 | |
| NM_001242390.1 | NP_001229319.1 | 6 - 7 | 613 | 54 | ||
| NM_001242391.1 | NP_001229320.1 | 6 - 7 | 613 | 54 | ||
| NM_134125.4 | NP_598886.1 | 6 - 7 | 613 | 54 | ||
| GenBank mRNA | AF502565.1 | - | 6 - 7 | 516 | 54 | |
| AK088909.1 | - | 6 - 7 | 563 | 54 | ||
| AK149902.1 | - | 6 - 7 | 585 | 54 | ||
| AY081142.1 | - | 6 - 7 | 564 | 54 | ||
| BC003249.1 | - | 6 - 7 | 615 | 3500118 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trip10
Genomic Map