Assay Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146180.1 | NP_001139652.1 | 8 - 9 | 1220 | 89 | |
| NM_144800.2 | NP_659049.2 | 9 - 10 | 1232 | 89 | ||
| GenBank mRNA | AK030533.1 | - | 7 - 8 | 626 | 89 | |
| AK129139.1 | - | 8 - 9 | 1217 | 89 | ||
| AY214918.1 | - | 9 - 10 | 733 | 89 | ||
| BC023682.1 | - | 3 - 4 | 338 | 89 | ||
| BC024131.1 | - | 9 - 10 | 1197 | 5101888 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtss1
Genomic Map