Assay Details
| Gene Symbol: Sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144907.1 | NP_659156.1 | 4 - 5 | 836 | 54 | |
| GenBank mRNA | AK143788.1 | - | 4 - 5 | 880 | 54 | |
| AK170547.1 | - | 880 | 54 | |||
| AK182914.1 | - | 2 - 3 | 256 | 54 | ||
| AK196889.1 | - | 1 - 2 | 65 | 54 | ||
| BC005672.1 | - | 4 - 5 | 836 | 3153129 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sesn2
Genomic Map