Assay Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291282.1 | NP_001278211.1 | 4 - 5 | 340 | 55 | |
| NM_145375.4 | NP_663350.2 | 4 - 5 | 848 | 55 | ||
| GenBank mRNA | AK046714.1 | - | 4 - 5 | 749 | 55 | |
| AK083132.1 | - | 4 - 5 | 851 | 55 | ||
| AK135100.1 | - | 4 - 5 | 830 | 55 | ||
| AK136581.1 | - | 4 - 5 | 341 | 55 | ||
| AK163621.1 | - | 4 - 5 | 773 | 55 | ||
| AK172109.1 | - | 4 - 5 | 764 | 55 | ||
| AK172145.1 | - | 4 - 5 | 749 | 55 | ||
| BC023123.1 | - | 4 - 5 | 730 | 3995280 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm6sf1
Genomic Map