Assay Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037937.3 | NP_001033026.1 | 4 - 5 | 607 | 65 | |
| NM_145470.3 | NP_663445.2 | 4 - 5 | 785 | 65 | ||
| GenBank mRNA | AK139888.1 | - | 4 - 5 | 607 | 65 | |
| AK145208.1 | - | 4 - 5 | 679 | 65 | ||
| AK220300.1 | - | 4 - 5 | 619 | 65 | ||
| BC004774.1 | - | 4 - 5 | 790 | 65 | ||
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK184788.1 | - | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Deptor
Genomic Map