Assay Details
| Gene Symbol: Rgs9bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145840.3 | NP_665839.1 | 1 | 514 | 67 | |
| GenBank mRNA | AF483907.1 | - | 1 | 152 | 67 | |
| AK044285.1 | - | 1 | 515 | 67 | ||
| AY229888.1 | - | 1 | 476 | 67 | ||
| BC118006.1 | - | 1 | 470 | 40081388 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050054 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134947 | AY229888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134947 | NM_145840.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134947 | AF483907.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25291 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136201 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134947 | AY229888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134947 | NM_145840.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134947 | AF483907.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P319165 | AK044285.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P319165 | AY229888.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P319165 | NM_145840.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25050028 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136201 | - | - |
| Whole Mouse Genome G4122A | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659470 | AY229888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659470 | NM_145840.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P319165 | AK044285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P319165 | AY229888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P319165 | NM_145840.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121728 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06442875 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06442876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06591530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050054 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25050065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25049995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25049999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25050053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25049994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06536132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06536138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06536139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06177024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25050050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562489 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562487 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440256_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562486 | - | - |
| Mouse Genome 430 2.0 Array | 1440256_at | - | - |
| Mouse Genome 430 2.0 Array | 1440256_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243923_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243923_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700025526.mm.1 | NM_145840.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025526.mm.1 | NM_145840.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025526.mm.1 | TC0700002855.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025525.mm.1 | NM_145840.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025525.mm.1 | NM_145840.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025525.mm.1 | TC0700002855.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025527.mm.1 | NM_145840.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025527.mm.1 | NM_145840.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025527.mm.1 | TC0700002855.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440256_PM_at | - | - |
| Mouse Expression Set 430 | 1440256_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440256_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440256_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 438851 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1690546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 840047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252964 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237501 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237501 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252964 | - | - |
| mouse-6 v1.1 (Illumina) | scl0243923.1_215-S | - | - |
| mouse-6 v1.1 (Illumina) | scl31468.1_378-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840047 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0243923.1_215-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31468.1_378-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840047 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237501 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2795473 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237501 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2795473 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31468.1_378-S | - | - |

Rgs9bp
Genomic Map