Assay Details
| Gene Symbol: Wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145940.2 | NP_666052.1 | 2 - 3 | 242 | 61 | |
| GenBank mRNA | AK031672.1 | - | 2 - 3 | 251 | 61 | |
| AK034907.1 | - | 2 - 3 | 286 | 61 | ||
| AK045131.1 | - | 2 - 3 | 251 | 61 | ||
| AK050495.1 | - | 2 - 3 | 250 | 61 | ||
| AK159608.1 | - | 2 - 3 | 271 | 61 | ||
| AK170563.1 | - | 2 - 3 | 286 | 61 | ||
| BC024811.1 | - | 2 - 3 | 210 | 61 | ||
| BC024883.1 | - | 2 - 3 | 206 | 61 | ||
| BC025560.1 | - | 2 - 3 | 242 | 5322999 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wipi1
Genomic Map