Assay Details
| Gene Symbol: Wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145940.2 | NP_666052.1 | 7 - 8 | 772 | 88 | |
| GenBank mRNA | AK031672.1 | - | 7 - 8 | 781 | 88 | |
| AK034907.1 | - | 7 - 8 | 816 | 88 | ||
| AK045131.1 | - | 7 - 8 | 782 | 88 | ||
| AK050495.1 | - | 7 - 8 | 780 | 88 | ||
| AK159608.1 | - | 7 - 8 | 801 | 88 | ||
| AK170563.1 | - | 7 - 8 | 816 | 88 | ||
| BC024811.1 | - | 7 - 8 | 740 | 88 | ||
| BC024883.1 | - | 7 - 8 | 736 | 88 | ||
| BC025560.1 | - | 7 - 8 | 772 | 5322999 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wipi1
Genomic Map