Assay Details
| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146041.2 | NP_666153.1 | 8 - 9 | 951 | 101 | |
| GenBank mRNA | BC031788.1 | - | 8 - 9 | 951 | 4165509 | 101 |
| BC093502.1 | - | 8 - 9 | 975 | 4215865 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmds
Genomic Map