Assay Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146198.2 | NP_666310.1 | 5 - 6 | 587 | 61 | |
| GenBank mRNA | AF518726.1 | - | 6 - 7 | 680 | 61 | |
| AK008219.1 | - | 5 - 6 | 641 | 61 | ||
| BC031742.1 | - | 5 - 6 | 588 | 4221537 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a11
Genomic Map