Assay Details
| Gene Symbol: Gemin8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146238.3 | NP_666350.2 | 2 - 3 | 114 | 76 | |
| GenBank mRNA | AK047511.1 | - | 3 - 4 | 209 | 76 | |
| AK053774.1 | - | 2 - 3 | 78 | 76 | ||
| AK160583.1 | - | 2 - 3 | 115 | 76 | ||
| BC023488.1 | - | 3 - 4 | 104 | 5010787 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047511.1, AK053774.1, AK160583.1, BC023488.1, NM_146238.3 transcripts.
Set Membership: |
|

Gemin8
Genomic Map