Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 2 - 3 | 179 | 83 | |
| GenBank mRNA | AK159780.1 | - | 2 - 3 | 106 | 83 | |
| AK216971.1 | - | 1 - 2 | 80 | 83 | ||
| BC018431.1 | - | 2 - 3 | 179 | 83 | ||
| BC031416.1 | - | 3 - 4 | 268 | 4012768 | 83 | |
| BM947393.1 | - | 2 - 3 | 91 | 83 | ||
| BY141405.1 | - | 2 - 3 | 196 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK159780.1,BC031416.1,NM_146248.2 transcripts.
Set Membership: |
|

Cchcr1
Genomic Map