Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 3 - 4 | 396 | 109 | |
| GenBank mRNA | AK159780.1 | - | 3 - 4 | 323 | 109 | |
| BC031416.1 | - | 4 - 5 | 485 | 4012768 | 109 | |
| BM947393.1 | - | 3 - 4 | 308 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cchcr1
Genomic Map