Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 16 - 17 | 2232 | 58 | |
| GenBank mRNA | AK159780.1 | - | 16 - 17 | 2159 | 58 | |
| BC031416.1 | - | 17 - 18 | 2321 | 4012768 | 58 | |
| BQ031569.1 | - | 2 - 3 | 58 | |||
| CJ238541.1 | - | 3 - 4 | 280 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cchcr1
Genomic Map