Assay Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153054.2 | NP_694694.1 | 14 - 15 | 1671 | 69 | |
| GenBank mRNA | AK076433.1 | - | 13 - 14 | 1396 | 69 | |
| AY779336.1 | - | 12 - 13 | 1329 | 69 | ||
| BC027192.1 | - | 14 - 15 | 1609 | 4239930 | 69 | |
| BC031146.1 | - | 14 - 15 | 1639 | 4974776 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a1
Genomic Map